Learn More
Electron Microscopy Sciences Omniprobe Lift-Out Grid 3-Post, Side Access, Cooper
SDP
Catalog No. 501902651
Encompass_Preferred
3 post copper lift-out grids, similar to 75964-01, in design but 35 micron thick with 1 edge lower for easy access. Packaged in glass vials
Material: Cooper
Catalog No. 50-190-2651 Supplier Electron Microscopy Sciences Supplier No. 7596407
3 post copper lift-out grids, similar to 75964-01, in design but 35 micron thick with 1 edge lower for easy access. Packaged in glass vials
Material: Cooper