Learn More
Electron Microscopy Sciences Omniprobe Lift-Out Grid 3-Post, Shallow Downset, Cooper
SDP
Catalog No. 501902652
Encompass_Preferred
3 post copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations with both vertical bar and "V" shaped attachment surfaces. These grids have a shallower downset and slightly wider center post than 75964-01. Packaged in glass vials.
Material: Copper
Catalog No. 50-190-2652 Supplier Electron Microscopy Sciences Supplier No. 7596408
3 post copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations with both vertical bar and "V" shaped attachment surfaces. These grids have a shallower downset and slightly wider center post than 75964-01. Packaged in glass vials.
Material: Copper