Learn More
Electron Microscopy Sciences Omniprobe Grid Storage Box
SDP
Catalog No. 5033770
Encompass
Storage box for 100 standard or haft grids - 3 mm diameter TEM grids. Box comes complete with base, lid and clips.
Catalog No. 50-337-70 Supplier Electron Microscopy Sciences Supplier No. 7596501
Storage box for 100 standard or haft grids - 3 mm diameter TEM grids. Box comes complete with base, lid and clips.