Learn More
Electron Microscopy Sciences Omniprobe In-Situ Probe Tip
SDP
Catalog No. 5033757
Encompass
Custom tungsten tip with a stainless steel shank, for use with AutoProbe 300, in-situ probe tip exchange systems and Short-Cut. Tip radius is 0.5um with 8 -10 degree taper angle.This tip is also Short-Cut compatible.
Catalog No. 50-337-57 Supplier Electron Microscopy Sciences Supplier No. 7596003
Custom tungsten tip with a stainless steel shank, for use with AutoProbe 300, in-situ probe tip exchange systems and Short-Cut. Tip radius is 0.5um with 8 -10 degree taper angle.This tip is also Short-Cut compatible.