Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ OmniPur™ Coomassie™ Blue R-250

Catalog No. 3345250GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-452-50GM 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-452-50GM Supplier MilliporeSigma™ Supplier No. 3345250GM

missing translation for 'validMainframeMsg'

Used for post-electrophoresis staining of proteins.

EMD Millipore OmniPur™ Coomassie Blue R-250 for applications requiring a high degree of sensitivity solubility.

  • Use for post-electrophoresis staining of proteins in applications requiring high degree of sensitivity or solubility
  • CAS: 6104-59-2
  • Molecular formula: C45H44N3NaO7S2
TRUSTED_SUSTAINABILITY

Specifications

Description Coomassie Blue R-250
Color Blue
Quantity 250 g
Product Type Protein Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.