Learn More
Description
- CAS: 2353-45-9
- Molecular formula: C37H34N2O10S3Na2
Specifications
Specifications
| Description | Crystalline Powder |
| Color | Green |
| Quantity | 50 g |
| Product Type | Protein Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.