Learn More
Electron Microscopy Sciences OMNISTOR CABINET BASE
SDP
Catalog No. 502413064
Encompass_Preferred
OmniStor Slide and Block Storage System is an economical and durable storage, made from corrosive protected steel. Its "EasyLock" system allows cabinetsto be stacked securely without removing drawers, fasteners or screw pates.
Catalog No. 50-241-3064 Supplier Electron Microscopy Sciences Supplier No. 6328167
OmniStor Slide and Block Storage System is an economical and durable storage, made from corrosive protected steel. Its "EasyLock" system allows cabinetsto be stacked securely without removing drawers, fasteners or screw pates.