Learn More
Sirius Automation Inc OmniTasker Benchtop Lab Robot
SDP
Catalog No. 5000078213
Encompass
The OmniTasker is a Large Capacity Modular Laboratory Robot for everyday tasks such as Powder Dispensing Analytical Weighing (4 5 or 6 place) Sample ID (1D or 2D barcodes) Vial Sorting Dilutions Extractions Internal Standard Addition Standard Prep Un-Capping Re-Capping Transfers Aliquoting Large Sample Capacity (max 32 microplate racks or custom equivalent) with choice of disposables and labware The software has easy to use interface (21CFR11 compliance options) with LIMS compatible data management The robot hardware is fast and precise and incorporates a built-in enclosure and optional hardware interlock to ensure safety Each robotic step is recorded and logged such that there is a unique and forensic sample history to be readily maintained
Catalog No. 50-000-78213 Supplier Sirius Automation Inc Supplier No. OMN60101
Add to Cart
Add to Cart
Request a Quote
The OmniTasker is a Large Capacity Modular Laboratory Robot for everyday tasks such as Powder Dispensing Analytical Weighing (4 5 or 6 place) Sample ID (1D or 2D barcodes) Vial Sorting Dilutions Extractions Internal Standard Addition Standard Prep Un-Capping Re-Capping Transfers Aliquoting Large Sample Capacity (max 32 microplate racks or custom equivalent) with choice of disposables and labware The software has easy to use interface (21CFR11 compliance options) with LIMS compatible data management The robot hardware is fast and precise and incorporates a built-in enclosure and optional hardware interlock to ensure safety Each robotic step is recorded and logged such that there is a unique and forensic sample history to be readily maintained