Learn More
Aviva Systems Biology Corp Omp Antibody - C-terminal region (ARP56669_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56669_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Omp. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: -ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-286-188