Learn More
Aviva Systems Biology Corp OMP Antibody - middle region (ARP56668_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56668_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OMP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OMP. Alternate names: -ApplicationWB. Species reactivity: Goat, Human, Mouse, Rat
Catalog No. 50-014-61946