Learn More
Biomatik Corporation Ompa Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11890
Encompass_Preferred
Ompa Polyclonal Antibody is a Salmonella typhi Polyclonal Antibody to ompA. The Ompa Polyclonal Antibody has been validated for the following applications: ELISA. Host: Rabbit. Species: Salmonella typhi. Immunogen: Recombinant Salmonella Typhi Outer Membrane Protein A Protein (27-349Aa). Applications: ELISA. Target Synonyms: ompA. Accession Number: Q8Z7S0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9233