Learn More
Biomatik Corporation Ompa Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11612
Encompass_Preferred
Ompa Polyclonal Antibody is a Haemophilus influenzae Polyclonal Antibody to ompA. The Ompa Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Haemophilus influenzae. Immunogen: Recombinant Haemophilus Influenzae Outer Membrane Protein P5 Protein (22-359Aa). Applications: ELISA, WB. Target Synonyms: ompA. Accession Number: P45996. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9238