Learn More
Biomatik Corporation Ompa Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11453
Encompass_Preferred
Ompa Polyclonal Antibody is a Rickettsia conorii Polyclonal Antibody to ompA. The Ompa Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Rickettsia conorii. Immunogen: Recombinant Rickettsia Conorii Outer Membrane Protein A Protein (1734-2021Aa). Applications: ELISA, WB. Target Synonyms: ompA. Accession Number: Q52657. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5664