Learn More
Biomatik Corporation Ompc Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11656
Encompass_Preferred
Ompc Polyclonal Antibody is a Escherichia coli O157:H7 Polyclonal Antibody to ompC. The Ompc Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli O157:H7. Immunogen: Recombinant Escherichia Coli O157:H7 Outer Membrane Protein C Protein (22-367Aa). Applications: ELISA, WB. Target Synonyms: ompC. Accession Number: Q8XE41. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9237