Learn More
Biomatik Corporation Ompf Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11714
Encompass_Preferred
Ompf Polyclonal Antibody is a Escherichia coli (strain K12) Polyclonal Antibody to OMPF. The Ompf Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli (strain K12). Immunogen: Recombinant Escherichia Coli (Strain K12) Ompf Protein (23-362Aa). Applications: ELISA, WB. Target Synonyms: OMPF. Accession Number: P02931. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9236