Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Omphalotin A | 186511-50-2 | 99.8% | 1318.73 | 1 MG
SDP

Catalog No. 5000399848
Encompass_Preferred

Omphalotin A is a cyclic peptide that exhibits nematicidal activity. It is isolated from microorganisms, specifically Omphalotus olearius.

  • Cyclic peptide with nematicidal activity
  • Purity: 99.8%
  • Molecular weight: 1318.73
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Microorganisms, Omphalotus olearius

Catalog No. 50-003-99848 Supplier Medchemexpress LLC Supplier No. HYP59571MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Omphalotin A is a cyclic peptide that exhibits nematicidal activity. It is isolated from microorganisms, specifically Omphalotus olearius.

  • Cyclic peptide with nematicidal activity
  • Purity: 99.8%
  • Molecular weight: 1318.73
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Microorganisms, Omphalotus olearius

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.