Learn More
Enzo Life Sciences clasto-Lactacystin beta-Lactone (100µg). CAS: 155975-72-7
SDP
Supplier:  Enzo Life Sciences BMLPI1080100
Encompass_Preferred
Highly specific, cell permeable, and irreversible 20S proteasome inhibitor. Inhibits ubiquitin proteasome pathway in cell culture (IC50=1µM). It has been suggested that the natural product lactacystin acts as a precursor for clasto-lactacystin β-lactone and that the latter is the sole species that interacts with the proteasome. Inhibits cathepsin A. Leads to a reduction of adipogenesis. Induces neurite growth and inhibits cell cycle progression.  Alternative name: Omuralide. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or acetonitrile. Long Term Storage: -20°C.
Catalog No. 50-201-1443