Learn More
Medchemexpress LLC Omzotirome | 1092551-88-6 | 99.9% | 328.41 | C19H24N2O3 | 100 MG
SDP
Catalog No. 5000240425
Encompass_Preferred
Omzotirome is a thyromimetic research compound used in preclinical and clinical studies of lipid metabolism and hyperlipidaemia. It is a functional analog of iodothyronines and is supplied with chemical and structural identifiers for laboratory research.
  • Thyromimetic activity useful for lipid metabolism studies.
  • Documented chemical identifiers (CAS 1092551-88-6, molecular formula C19H24N2O3).
  • High reported purity suitable for analytical and biological assays.
  • Available in multiple small pack sizes for research use.
  • Provided with structural data and registry cross-references for traceability.
Catalog No. 50-002-40425 Supplier Medchemexpress LLC Supplier No. HY106425100MG
Omzotirome is a thyromimetic research compound used in preclinical and clinical studies of lipid metabolism and hyperlipidaemia. It is a functional analog of iodothyronines and is supplied with chemical and structural identifiers for laboratory research.
  • Thyromimetic activity useful for lipid metabolism studies.
  • Documented chemical identifiers (CAS 1092551-88-6, molecular formula C19H24N2O3).
  • High reported purity suitable for analytical and biological assays.
  • Available in multiple small pack sizes for research use.
  • Provided with structural data and registry cross-references for traceability.