Learn More
Medchemexpress LLC Omzotirome | 1092551-88-6 | 99.9% | 328.41 g·mol^-1 | C19H24N2O3 | 50 MG
SDP
Catalog No. 5000240922
Encompass_Preferred
Omzotirome is a thyromimetic research compound, a functional analog of iodothyronines, used in preclinical studies of lipid metabolism and hyperlipidaemia. It acts as a metabolic modulator that increases energy expenditure and fatty acid oxidation, and is supplied at high chemical purity for laboratory research.
  • Used in hyperlipidaemia and metabolic research.
  • Acts as a thyromimetic metabolic modulator.
  • Increases energy expenditure and fatty acid oxidation in preclinical models.
  • Provided in small-scale packaging suitable for bench research.
  • Solid form recommended to be stored at 4°C; solutions should be stored frozen.
Catalog No. 50-002-40922 Supplier Medchemexpress LLC Supplier No. HY10642550MG
Omzotirome is a thyromimetic research compound, a functional analog of iodothyronines, used in preclinical studies of lipid metabolism and hyperlipidaemia. It acts as a metabolic modulator that increases energy expenditure and fatty acid oxidation, and is supplied at high chemical purity for laboratory research.
  • Used in hyperlipidaemia and metabolic research.
  • Acts as a thyromimetic metabolic modulator.
  • Increases energy expenditure and fatty acid oxidation in preclinical models.
  • Provided in small-scale packaging suitable for bench research.
  • Solid form recommended to be stored at 4°C; solutions should be stored frozen.