Learn More
Medchemexpress LLC HY-112822 10mg Medchemexpress, ON-013100 CAS:865783-95-5 Purity:>98%
SDP
Catalog No. 501969184
Encompass_Preferred
Medchemexpress, HY-112822 10mg ON-013100 CAS:865783-95-5 ON-013100, an antineoplastic drug, acts a mitotic inhibitor that could inhibit Cyclin D1 expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9184 Supplier Medchemexpress LLC Supplier No. HY11282210MG
Medchemexpress, HY-112822 10mg ON-013100 CAS:865783-95-5 ON-013100, an antineoplastic drug, acts a mitotic inhibitor that could inhibit Cyclin D1 expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.