Learn More
Cayman Chemical ON-01910 (sodium salt), Purity: ≥98%, CAS: 1225497-78-8, 25 mg
SDP
Catalog No. 502971160
Encompass_Preferred
A potent, non-ATP-competitive inhibitor of Plk1 (IC50 = 9 nM); induces apoptosis in a wide array of tumor cell lines (GI50 = 50-200 nM); active in vivo when administered intraperitoneally and synergizes with chemotherapeutic agents
Catalog No. 50-297-1160 Supplier Cayman Chemical Supplier No. 1555325 MG
A potent, non-ATP-competitive inhibitor of Plk1 (IC50 = 9 nM); induces apoptosis in a wide array of tumor cell lines (GI50 = 50-200 nM); active in vivo when administered intraperitoneally and synergizes with chemotherapeutic agents