Learn More
Cayman Chemical ONO-4059 (hydrochloride), Purity: ≥98%, CAS: 1439901-97-9, 50 mg
SDP
Catalog No. 502950411
Encompass_Preferred
A BTK inhibitor (IC50 = 2.2 nM); selective for BTK over LCK, LYN, and Fyn at 1 µM; inhibits B cell proliferation and activation in vitro and reduces tumor growth in a TMD8 mouse xenograft model
Catalog No. 50-295-0411 Supplier Cayman Chemical Supplier No. 3281850 MG
A BTK inhibitor (IC50 = 2.2 nM); selective for BTK over LCK, LYN, and Fyn at 1 µM; inhibits B cell proliferation and activation in vitro and reduces tumor growth in a TMD8 mouse xenograft model