Learn More
Cayman Chemical on-7300243 10mg
SDP
Catalog No. 502987713
Encompass_Preferred
An LPA1 receptor antagonist (IC50 = 0.16 µM); selective LPA1 over the LPA2 and LPA3 receptors (IC50s = 8.6 and >10 µM, respectively); reduces LPA-induced intraurethral pressure induced by LPA in rats (ID50 = 11.6 mg/kg)
Catalog No. 50-298-7713 Supplier Cayman Chemical Supplier No. 2205210 MG
An LPA1 receptor antagonist (IC50 = 0.16 µM); selective LPA1 over the LPA2 and LPA3 receptors (IC50s = 8.6 and >10 µM, respectively); reduces LPA-induced intraurethral pressure induced by LPA in rats (ID50 = 11.6 mg/kg)