Learn More
Cayman Chemical on-8711dIcyclohexylamIn s 10mg
SDP
Catalog No. 502916680
Encompass_Preferred
An EP1 receptor antagonist (Kis = 0.6 and 1.7 nM for human and mouse receptors, respectively, expressed in CHO cells); is selective for EP1 over EP2, EP3, and EP4 (Kis = >1,000, 67, and >1,000 nM, respectively, for mouse receptors expressed in CHO cells); inhibits PGE2-induced increases in intracellular calcium concentrations in cells expressing human or mouse EP1 (EC50s = 50 and 210 nM, respectively); reduces the incidence of squamous cell carcinoma and severe dysplasia induced by 4-nitroquinoline 1-oxide in a rat model of tongue carcinogenesis at 400 and 800 ppm in the diet
Catalog No. 50-291-6680 Supplier Cayman Chemical Supplier No. 3843210 MG
An EP1 receptor antagonist (Kis = 0.6 and 1.7 nM for human and mouse receptors, respectively, expressed in CHO cells); is selective for EP1 over EP2, EP3, and EP4 (Kis = >1,000, 67, and >1,000 nM, respectively, for mouse receptors expressed in CHO cells); inhibits PGE2-induced increases in intracellular calcium concentrations in cells expressing human or mouse EP1 (EC50s = 50 and 210 nM, respectively); reduces the incidence of squamous cell carcinoma and severe dysplasia induced by 4-nitroquinoline 1-oxide in a rat model of tongue carcinogenesis at 400 and 800 ppm in the diet