Learn More
Cayman Chemical on-AE3-208 10mg
SDP
Catalog No. 502970033
Encompass_Preferred
An antagonist of the EP4 receptor (Ki = 1.3 nM) which less potently affects EP3, FP, and TP receptors (Kis = 30, 790, and 2,400 nM, respectively) and is without effect on other prostanoid receptors; implicates EP4 signaling in immune and autoimmune responses, inflammation, and cancer
Catalog No. 50-297-0033 Supplier Cayman Chemical Supplier No. 1452210 MG
An antagonist of the EP4 receptor (Ki = 1.3 nM) which less potently affects EP3, FP, and TP receptors (Kis = 30, 790, and 2,400 nM, respectively) and is without effect on other prostanoid receptors; implicates EP4 signaling in immune and autoimmune responses, inflammation, and cancer