Learn More
MSC Ideal Warehouse Innovations Inc. Propane Gloves, Universal, PVC, Foam-Insulated, 1 Pair
Catalog No. 5000687171
Encompass
These general purpose chemical-resistant gloves are designed to protect hands against liquid propane burns and frostbite during propane cylinder changes. Constructed with a high-visibility orange polyvinylchloride coating and a foam-insulated lining, these unsupported gloves are engineered for durability and safety in challenging environments.
  • Provides protection against liquid propane burns and frostbite
  • Features a foam-insulated lining for comfort and thermal protection
  • Works in a temperature range of -4°F to 175°F
  • Designed with a sandy grip surface for improved handling
  • Includes an attached chain for mounting to an overhead guard for easy access
  • Silicone-free construction
  • High-visibility orange color for increased awareness
Catalog No. 50-006-87171 Supplier MSC Supplier No. 04206538
These general purpose chemical-resistant gloves are designed to protect hands against liquid propane burns and frostbite during propane cylinder changes. Constructed with a high-visibility orange polyvinylchloride coating and a foam-insulated lining, these unsupported gloves are engineered for durability and safety in challenging environments.
  • Provides protection against liquid propane burns and frostbite
  • Features a foam-insulated lining for comfort and thermal protection
  • Works in a temperature range of -4°F to 175°F
  • Designed with a sandy grip surface for improved handling
  • Includes an attached chain for mounting to an overhead guard for easy access
  • Silicone-free construction
  • High-visibility orange color for increased awareness