Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED ON-TARGETplus Human TRIM63 siRNA, SMARTPool, 5nmol
Catalog No. NC3233853
Encompass
This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is an E3 ubiquitin ligase that localizes to the Z-line and M-line lattices of myofibrils. This protein plays an important role in the atrophy of skeletal and cardiac muscle and is required for the degradation of myosin heavy chain proteins, myosin light chain, myosin binding protein, and for muscle-type creatine kinase. [provided by RefSeq, Feb 2012].
Catalog No. NC3233853 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. L007093000005
Add to Cart
Add to Cart
This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is an E3 ubiquitin ligase that localizes to the Z-line and M-line lattices of myofibrils. This protein plays an important role in the atrophy of skeletal and cardiac muscle and is required for the degradation of myosin heavy chain proteins, myosin light chain, myosin binding protein, and for muscle-type creatine kinase. [provided by RefSeq, Feb 2012].