Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED ON-TARGETplus siRNA Reagents - Human, MAP2K5, SMARTPool, 5 nmol
Catalog No. NC1855325
Encompass
A mixture of 4 siRNA provided as a single reagent, providing advantages in both potency and specificity.
Catalog No. NC1855325 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. L003966000005
A mixture of 4 siRNA provided as a single reagent, providing advantages in both potency and specificity.