Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Dharmacon™ ON-TARGETplus siRNA, Human, CDK6, SMARTpool, 1 nmol
Catalog No. NC3931306
Encompass
This SMARTpool reagent provides a mixture of four siRNAs designed for high-specificity human CDK6 gene silencing. Utilizing patented chemical modifications, this reagent effectively reduces off-target activity by up to 90% compared to unmodified siRNA, ensuring reliable and reproducible RNAi experimental results.
  • Features patented chemical modifications to minimize off-target effects
  • Supplied as a SMARTpool of four individual siRNAs for enhanced potency and specificity
  • Guaranteed gene silencing performance
  • Validated for use in gene knockdown and RNAi experiments
  • Routinely utilized at 5 to 25 nM concentrations
  • Compatible with human transcript isoform coverage
Catalog No. NC3931306 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. L003240000010
This SMARTpool reagent provides a mixture of four siRNAs designed for high-specificity human CDK6 gene silencing. Utilizing patented chemical modifications, this reagent effectively reduces off-target activity by up to 90% compared to unmodified siRNA, ensuring reliable and reproducible RNAi experimental results.
  • Features patented chemical modifications to minimize off-target effects
  • Supplied as a SMARTpool of four individual siRNAs for enhanced potency and specificity
  • Guaranteed gene silencing performance
  • Validated for use in gene knockdown and RNAi experiments
  • Routinely utilized at 5 to 25 nM concentrations
  • Compatible with human transcript isoform coverage