Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Dharmacon™ ON-TARGETplus siRNA, Human WIZ, SMARTpool, 5 nmol
Catalog No. NC3911767
Encompass
This product provides highly specific, reliable gene silencing reagents designed to minimize off-target effects through patented chemical modifications. Supplied in a SMARTpool format, which consists of a mixture of four siRNAs, this reagent maximizes potency while reducing off-target activity by up to 90% compared to unmodified sequences.
  • Features patented modifications to significantly reduce off-target effects
  • Provides reliable gene silencing for experimental knockdown applications
  • Includes four distinct siRNAs in a single reagent mixture for enhanced potency
  • Compatible with concentrations ranging from 5 to 25 nM
  • Off-target effects reduced by up to 90% compared to unmodified alternatives
  • Supports gene silencing in human, mouse, and rat species
Catalog No. NC3911767 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. L031240010005
This product provides highly specific, reliable gene silencing reagents designed to minimize off-target effects through patented chemical modifications. Supplied in a SMARTpool format, which consists of a mixture of four siRNAs, this reagent maximizes potency while reducing off-target activity by up to 90% compared to unmodified sequences.
  • Features patented modifications to significantly reduce off-target effects
  • Provides reliable gene silencing for experimental knockdown applications
  • Includes four distinct siRNAs in a single reagent mixture for enhanced potency
  • Compatible with concentrations ranging from 5 to 25 nM
  • Off-target effects reduced by up to 90% compared to unmodified alternatives
  • Supports gene silencing in human, mouse, and rat species