Learn More
Medchemexpress LLC HY-100789 5mg , ON1231320 CAS:1312471-39-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1007895MG
Encompass_Preferred
HY-100789 5mg ON1231320 CAS: 1312471-39-8 ON1231320 is a highly specific polo like kinase 2 (PLK2) inhibitor with an IC50 of 0.31 µM. ON1231320 blocks tumor cell cycle progression in the G2/M phase in mitosis, causing apoptotic cell death. ON1231320, an arylsulfonyl pyrido-pyrimidinone, has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1241