Learn More
Selleck Chemical LLC ON123300 5mg 1357470-29-1
SDP
Supplier:  Selleck Chemical LLC S81615MG
Encompass_Preferred
ON123300 is a potent and multi-targeted kinase inhibitor with IC50 of 3.9 nM, 5 nM, 26 nM, 26 nM, 9.2 nM and 11nM for CDK4, Ark5/NUAK1, PDGFR?, FGFR1, RET (c-RET), and Fyn, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5729