Learn More
Medchemexpress LLC HY-14463 10mg Medchemexpress, Onalespib CAS:912999-49-6 Purity:>98%
SDP
Catalog No. 502029052
Encompass_Preferred
Medchemexpress, HY-14463 10mg Onalespib CAS:912999-49-6 Onalespib (AT13387) is a long-acting second-generation Hsp90 inhibitor with a Kd of 0.71 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9052 Supplier Medchemexpress LLC Supplier No. HY1446310MG
Medchemexpress, HY-14463 10mg Onalespib CAS:912999-49-6 Onalespib (AT13387) is a long-acting second-generation Hsp90 inhibitor with a Kd of 0.71 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.