Learn More
Selleck Chemical LLC Onatasertib
SDP
Catalog No. 5000003317
Encompass_Preferred
Onatasertib (CC 223) is a potent selective and orally bioavailable mTOR inhibitor with IC50 of 16 nM 200-fold selectivity over the related PI3K- Phase 1/2
Catalog No. 50-000-03317 Supplier Selleck Chemical LLC Supplier No. S78865MG
Onatasertib (CC 223) is a potent selective and orally bioavailable mTOR inhibitor with IC50 of 16 nM 200-fold selectivity over the related PI3K- Phase 1/2