Learn More
Medchemexpress LLC HY-111343 5mg Medchemexpress, ONC212 CAS:1807861-48-8 Purity:>98%
SDP
Catalog No. 501968835
Encompass_Preferred
Medchemexpress, HY-111343 5mg ONC212 CAS:1807861-48-8 ONC212, a fluorinated-ONC201 analogue, is a promising anti-cancer drug and also a selective agonist of GPR132. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8835 Supplier Medchemexpress LLC Supplier No. HY1113435MG
Medchemexpress, HY-111343 5mg ONC212 CAS:1807861-48-8 ONC212, a fluorinated-ONC201 analogue, is a promising anti-cancer drug and also a selective agonist of GPR132. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.