Learn More
Medchemexpress LLC Oncaspar | 130167-69-0 | 99.9% | 1 MG
SDP
Catalog No. 5000346108
Encompass_Preferred
Oncaspar (PEG-L-asparaginase) is a pegylated form of L-asparaginase derived from Escherichia coli. It functions by breaking down asparagine in the bloodstream, playing a crucial role in acute lymphoblastic leukemia (ALL) research through asparagine hydrolysis and activation of the integrated stress response (ISR) pathway.
  • Pegylated form of L-asparaginase.
  • Breaks down asparagine in the bloodstream.
  • Important for acute lymphoblastic leukemia (ALL) research.
  • Suitable for ELISA, FACS, and functional assays.
  • Can be reconstituted or diluted with sterile PBS or saline.
  • Appears as a colorless to light yellow liquid.
Catalog No. 50-003-46108 Supplier Medchemexpress LLC Supplier No. HY1088605ML
Oncaspar (PEG-L-asparaginase) is a pegylated form of L-asparaginase derived from Escherichia coli. It functions by breaking down asparagine in the bloodstream, playing a crucial role in acute lymphoblastic leukemia (ALL) research through asparagine hydrolysis and activation of the integrated stress response (ISR) pathway.
  • Pegylated form of L-asparaginase.
  • Breaks down asparagine in the bloodstream.
  • Important for acute lymphoblastic leukemia (ALL) research.
  • Suitable for ELISA, FACS, and functional assays.
  • Can be reconstituted or diluted with sterile PBS or saline.
  • Appears as a colorless to light yellow liquid.