Learn More
ACROBiosystems Onchocerca volvulus OV16 protein, GST Tag
Catalog No. 502055019
Encompass_Preferred
Recombinant Protein;1MG;Onchocerca volvulus OV16, GST Tag (OV6-V51G4) is expressed from E.coli cells. It contains AA Lys 17 - Asp 197 (Accession # P31729-1).;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 46.8 kDa. The protein migrates as 45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5019 Supplier ACROBiosystems Supplier No. OV6V51G41MG
Recombinant Protein;1MG;Onchocerca volvulus OV16, GST Tag (OV6-V51G4) is expressed from E.coli cells. It contains AA Lys 17 - Asp 197 (Accession # P31729-1).;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 46.8 kDa. The protein migrates as 45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.