Learn More
Ion Torrent™ Oncomine™ Comprehensive Assay Plus, automated library preparation
Specifications
Specifications
| Chip Compatibility | Ion 550, Ion 560 |
| Content And Storage | Store at -30°C to -10°C. |
| Sample Type | FFPE Tissue, RNA |
| Sequencing Type | Next Generation Sequencing |
| For Use With (Equipment) | Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef |
| For Use With (Application) | Oncology Research |
| Libraries | Amplicon |
| Nucleic Acid Type | DNA and RNA |
| No. of Samples | Up to 32 Samples |
| Product Line | Oncomine |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.