Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Oncomine™ Comprehensive Assay Plus RNA, manual library preparation

Catalog No. A48578
Encompass_Preferred
Change view
Click to view available options
Quantity:
24 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A48578 24 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A48578 Supplier Ion Torrent™ Supplier No. A48578
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Oncomine Comprehensive Assay Plus RNA contains a targeted, multi-biomarker panel that enables highly sensitive and robust detection of known and novel fusions covering >1300 isoforms from 49 fusion drivers associated with solid tumors.

Key features of the assay include:

  • Detection of targeted known-fusion detection, non-targeted detection of novel fusions, and novel fusion detection with unknown partners
  • Highly sensitive and robust methodology with the ability to detect tiny levels of a tumor-specific fusion transcript in background normal RNA
  • Robust performance from as little as 10 ng per pool of RNA isolated from FFPE samples including fine needle biopsies
  • Characterized with molecular standards and controls
  • Content driven by the Oncomine Knowledgebase and experienced scientists helps assure coverage of key targets aligned to published evidence

The Oncomine Comprehensive Assay Plus RNA provides the reagents for library construction and two pools of multiplex PCR primers for preparation of amplicon libraries from formalin-fixed paraffin embedded (FFPE) tumor samples. The assay is optimized for sequencing on the Ion GeneStudio S5 systems with the Ion 550 Chip to enable up to 32 samples and a no-template control (NTC) per run.

When combined with Oncomine Reporter Software, you can focus on key drivers of cancer in your research.

TRUSTED_SUSTAINABILITY

Specifications

Chip Compatibility Ion 550, Ion 560
Content And Storage Store at -30°C to -10°C.
Sample Type FFPE Tissue
Sequencing Type Next Generation Sequencing
For Use With (Equipment) Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
For Use With (Application) Oncology Research
Libraries Amplicon
Nucleic Acid Type RNA
No. of Samples Up to 24 Samples
Product Line Oncomine
Product Type Assay
Quantity 24 Samples
Throughput Up to 32 Samples per Ion 550 Chip
Workflow Step Panel and Library
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.