Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Oncomine™ Comprehensive Assay v3 GX

Catalog No. A46296
Encompass_Preferred
Change view
Click to view available options
Quantity:
16 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A46296 16 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A46296 Supplier Ion Torrent™ Supplier No. A46296
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Specifications

Chip Compatibility GX5
Content And Storage Store at -30°C to -10°C.
Sample Type FFPE Tissue, RNA
Sequencing Type Next Generation Sequencing
For Use With (Equipment) Genexus Integrated Sequencer
For Use With (Application) Oncology Research
Libraries Amplicon
Nucleic Acid Type DNA and RNA
No. of Samples Up to 16 Samples
Product Line Oncomine
Product Type Assay
Quantity 16 Samples
Shipping Condition Approved for shipment on Wet or Dry Ice
Throughput Up to 6 Samples per GX5 Chip
Workflow Step Panel and Library
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.