Learn More
Ion Torrent™ Oncomine™ Dx Express Test FFPE DNA and RNA Control Kit
Description
The Ion Torrent Oncomine Dx Express Test FFPE DNA and RNA Control Kit contains DNA, RNA, and no-template controls for use with the Oncomine Dx Express Test.
The Oncomine Dx Express Test is an in vitro diagnostic assay that uses targeted next-generation sequencing (NGS) to detect biomarkers across 46 genes. The Oncomine™ Dx Express Test is a qualitative in vitro diagnostic test that uses targeted next generation sequencing (NGS) technology to detect substitutions, insertions, and deletions in 42 genes from DNA, copy number variants (CNVs) in 10 genes from DNA, and fusions or splice variants in 18 genes from RNA isolated from formalin-fixed, paraffin-embedded (FFPE) tumor samples using the Genexus™ Dx Integrated Sequencer.
The test is designed for use with the Genexus Dx System. Its fully automated workflow—from sample preparation to results—can be completed in as little 24 hours, supporting timely decision-making in clinical practice.
Specifications
Specifications
| Chip Compatibility | Genexus GX5 Dx |
| Content And Storage | Store at -30°C to -10°C. |
| Sample Type | Free DNA, RNA Control |
| Sequencing Type | Next Generation Sequencing |
| For Use With (Equipment) | Genexus Dx Integrated Sequencer |
| For Use With (Application) | Oncology Diagnostics |
| Libraries | Amplicon |
| No. of Reactions | 12 Reactions (4 Runs) |
| Nucleic Acid Type | DNA and RNA |
| No. of Samples | Up to 56 FFPE Samples |
| Show More |
For In Vitro Diagnostic Use. Not available in all countries or regions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.