Learn More
Ion Torrent™ Oncomine™ Dx Express Test Panel
Description
The Oncomine Dx Express Test is an in vitro diagnostic assay that uses targeted next-generation sequencing (NGS) to detect biomarkers across 46 genes. The Oncomine™ Dx Express Test is a qualitative in vitro diagnostic test that uses targeted next generation sequencing (NGS) technology to detect substitutions, insertions, and deletions in 42 genes from DNA, copy number variants (CNVs) in 10 genes from DNA, and fusions or splice variants in 18 genes from RNA isolated from formalin-fixed, paraffin-embedded (FFPE) tumor samples using the Genexus™ Dx Integrated Sequencer.
The test is designed for use with the Genexus Dx System. Its fully automated workflow—from sample preparation to results—can be completed in as little 24 hours, supporting timely decision-making in clinical practice.
Additional products that are required to complete the Oncomine Dx Express Test workflow include:
- Genexus Dx Integrated Sequencer
- Genexus Dx Library Strips 1 and 2-HD
- Genexus Dx Templating Strips 3-GX5 and 4
- Genexus Dx Sequencing Kit
- Genexus Dx Barcodes 1-32 HD
- Genexus Dx GX5 Chip and Genexus Coupler
- Genexus Dx Pipette Tips
- Oncomine Express Test FFPE DNA and RNA Control Kit
- Oncomine Express Test Plasma cfTNA Control Kit*
- MicroAmp EnduraPlate Optical 96-Well Clear GPLE Reaction Plates with Barcode
- Thermo Scientific Adhesive PCR Plate Foils
*Not available in the U.S.
Specifications
Specifications
| Chip Compatibility | Genexus GX5 Dx |
| Content And Storage | Store at -30°C to -10°C. |
| Sample Type | FFPE Tumor Tissue, Plasma* |
| Sequencing Type | Next Generation Sequencing |
| For Use With (Equipment) | Genexus Dx Integrated Sequencer |
| For Use With (Application) | Oncology Diagnostics |
| Libraries | Amplicon |
| No. of Reactions | 32 Reactions |
| Nucleic Acid Type | DNA, RNA, or cfTNA |
| No. of Samples | Up to 14 FFPE or 20 Plasma Samples |
| Show More |
For In Vitro Diagnostic Use. Not available in all countries or regions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.