Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oncomine™ Myeloid Research Assay—Chef Ready

Catalog No. A36941
Encompass_Preferred
Change view
Click to view available options
Quantity:
32 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A36941 32 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A36941 Supplier LIFE TECHNOLOGIES Supplier No. A36941
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Sample Type DNA
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) Ion PGM™ System
Libraries Targeted Sequencing Library
No. of Reactions 32
Product Line Oncomine
Quantity 32 reactions
Shipping Condition Dry Ice
Workflow Step DNA Extraction
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.