Learn More
Oncomine™ Myeloid Research Assay
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C). |
| Sample Type | DNA |
| Sequencing Type | Genome and DNA Sequencing |
| For Use With (Equipment) | Ion PGM™ System |
| Libraries | Targeted Sequencing Library |
| No. of Reactions | 24 |
| Product Line | Oncomine |
| Quantity | 24 reactions |
| Shipping Condition | Dry Ice |
| Workflow Step | DNA Extraction |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.