Learn More
Biocare Medical, LLC Biocare Medical ONCORE Pro Rabbit HRP Detection, High Sensitivity, Biotin-Free, 60 Tests
SDP
Catalog No. NC2962817
Encompass_Preferred
The Biocare Medical ONCORE Pro Rabbit HRP Detection is a highly sensitive detection system designed for immunohistochemistry (IHC) that targets rabbit IgG antibodies. Its proprietary micropolymer technology minimizes non-specific staining while ensuring clear results. Ideal for formalin-fixed, paraffin-embedded (FFPE) tissues, this ready-to-use product enhances the ease of use for automated staining protocols, setting it apart from biotin-based systems.
Catalog No. NC2962817 Supplier Biocare Medical, LLC Supplier No. OPRI6008T60
The Biocare Medical ONCORE Pro Rabbit HRP Detection is a highly sensitive detection system designed for immunohistochemistry (IHC) that targets rabbit IgG antibodies. Its proprietary micropolymer technology minimizes non-specific staining while ensuring clear results. Ideal for formalin-fixed, paraffin-embedded (FFPE) tissues, this ready-to-use product enhances the ease of use for automated staining protocols, setting it apart from biotin-based systems.