Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OncoScan™ CNV Assay for Research

Catalog No. 902695
Encompass_Preferred
Change view
Click to view available options
Quantity:
24 samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-269-5 24 samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-269-5 Supplier LIFE TECHNOLOGIES Supplier No. 902695
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Type CNV Assay
Array Cytogenetics, Copy Number
Number of Arrays 48 arrays
Format Genechip Probe Array
Content And Storage • OncoScan CNV Array (Cat. No. 902694)
• OncoScan CNV Reagent Kit (Cat. No. 902692TS)

Includes OncoScan CNV Array (Cat. No. 902694), OncoScan CNV Reagent Kit (Cat. No. 902692TS)
No. of Samples 24 samples
Number of Markers over 220,000 SNPs
Quantity 24 samples

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.