Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oncostatin M | 106956-32-5 | >95.0% | 50 UG
SDP

Catalog No. 5000291601
Encompass_Preferred

Recombinant human Oncostatin M (OSM), expressed in HEK293 cells and supplied as a tag-free, lyophilized powder. The glycosylated cytokine is intended for in vitro cell proliferation and signaling studies; typical activity is ED50 0.2-1.2 ng/mL (TF-1 assay), with endotoxin <1 EU/μg and purity >95%.

  • Recombinant human protein expressed in HEK293 cells.
  • Tag-free, lyophilized powder for long-term storage.
  • Glycosylated; appears ~28-32 kDa on SDS-PAGE.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Low endotoxin level: <1 EU/μg (LAL).
  • Biological activity: ED50 0.2-1.2 ng/mL in TF-1 proliferation assay.
  • Recommend reconstitution ≥100 μg/mL and store aliquots at -20 °C/-80 °C.

Catalog No. 50-002-91601 Supplier Medchemexpress LLC Supplier No. HYP7466250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Oncostatin M (OSM), expressed in HEK293 cells and supplied as a tag-free, lyophilized powder. The glycosylated cytokine is intended for in vitro cell proliferation and signaling studies; typical activity is ED50 0.2-1.2 ng/mL (TF-1 assay), with endotoxin <1 EU/μg and purity >95%.

  • Recombinant human protein expressed in HEK293 cells.
  • Tag-free, lyophilized powder for long-term storage.
  • Glycosylated; appears ~28-32 kDa on SDS-PAGE.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Low endotoxin level: <1 EU/μg (LAL).
  • Biological activity: ED50 0.2-1.2 ng/mL in TF-1 proliferation assay.
  • Recommend reconstitution ≥100 μg/mL and store aliquots at -20 °C/-80 °C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.