Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oncostatin M (human, recombinant) | 106956-32-5 | >95.0% | 10 UG
SDP

Catalog No. 5000291598
Encompass_Preferred

Recombinant human Oncostatin M (OSM) produced in HEK293 cells. This lyophilized, tag-free cytokine is provided for research use; it modulates cytokine production (for example, IL-6, G-CSF, GM-CSF), inhibits certain tumor cell lines, and stimulates Kaposi's sarcoma cell proliferation. Supplied with high purity and recommended frozen storage.

  • Expression system: HEK293 cells
  • Tag free, lyophilized preparation
  • Purity greater than 95% by reducing SDS-PAGE
  • Molecular weight approximately 28-32 kDa (glycosylated)
  • Sequence accession P13725 (A26-R221)
  • Do not reconstitute below 100 μg/mL in ddH2O
  • Store at -20°C; aliquot and freeze at -80°C for extended storage

Catalog No. 50-002-91598 Supplier Medchemexpress LLC Supplier No. HYP7466210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Oncostatin M (OSM) produced in HEK293 cells. This lyophilized, tag-free cytokine is provided for research use; it modulates cytokine production (for example, IL-6, G-CSF, GM-CSF), inhibits certain tumor cell lines, and stimulates Kaposi's sarcoma cell proliferation. Supplied with high purity and recommended frozen storage.

  • Expression system: HEK293 cells
  • Tag free, lyophilized preparation
  • Purity greater than 95% by reducing SDS-PAGE
  • Molecular weight approximately 28-32 kDa (glycosylated)
  • Sequence accession P13725 (A26-R221)
  • Do not reconstitute below 100 μg/mL in ddH2O
  • Store at -20°C; aliquot and freeze at -80°C for extended storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.