Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oncostatin M (human), recombinant (HEK293) | 106956-32-5 | >95.0% | 100 UG
SDP

Catalog No. 5000293196
Encompass_Preferred

Recombinant human Oncostatin M (OSM) expressed in HEK293 mammalian cells and provided lyophilized for research use. The product corresponds to human residues A26-R221, is supplied tag-free, and is intended for cell biology and biochemical assays that require a glycosylated, mammalian-expressed cytokine. Store at -20 °C and follow manufacturer instructions for reconstitution and handling.

  • Recombinant human Oncostatin M (A26-R221) produced in HEK293 cells.
  • Tag free preparation for downstream assays.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Glycosylated protein with apparent mass approximately 28-32 kDa.
  • Lyophilized formulation with stabilizers to support stability.
  • Store at -20 °C; stable for two years when stored as recommended.

Catalog No. 50-002-93196 Supplier Medchemexpress LLC Supplier No. HYP74662100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Oncostatin M (OSM) expressed in HEK293 mammalian cells and provided lyophilized for research use. The product corresponds to human residues A26-R221, is supplied tag-free, and is intended for cell biology and biochemical assays that require a glycosylated, mammalian-expressed cytokine. Store at -20 °C and follow manufacturer instructions for reconstitution and handling.

  • Recombinant human Oncostatin M (A26-R221) produced in HEK293 cells.
  • Tag free preparation for downstream assays.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Glycosylated protein with apparent mass approximately 28-32 kDa.
  • Lyophilized formulation with stabilizers to support stability.
  • Store at -20 °C; stable for two years when stored as recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.