Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oncostatin M (human recombinant, HEK293) | 106956-32-5 | >95.0% | 20 UG
SDP

Catalog No. 5000291599
Encompass_Preferred

Recombinant human Oncostatin M produced in HEK293 cells and supplied lyophilized as a tag-free protein corresponding to residues A26-R221 (UniProt P13725). Designed for biochemical and cell-based research that requires biologically active human OSM.

  • Recombinant human Oncostatin M, residues A26-R221.
  • Expression host: HEK293 (human embryonic kidney cells).
  • Tag free preparation for native-sequence studies.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass 22.2 kDa; observed ~28-32 kDa due to glycosylation.
  • Lyophilized formulation; refer to lot-specific COA for buffer details.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O; aliquot and store at -20°C or -80°C for long term.

Catalog No. 50-002-91599 Supplier Medchemexpress LLC Supplier No. HYP7466220UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Oncostatin M produced in HEK293 cells and supplied lyophilized as a tag-free protein corresponding to residues A26-R221 (UniProt P13725). Designed for biochemical and cell-based research that requires biologically active human OSM.

  • Recombinant human Oncostatin M, residues A26-R221.
  • Expression host: HEK293 (human embryonic kidney cells).
  • Tag free preparation for native-sequence studies.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass 22.2 kDa; observed ~28-32 kDa due to glycosylation.
  • Lyophilized formulation; refer to lot-specific COA for buffer details.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O; aliquot and store at -20°C or -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.